Skip to Content
Merck
All Photos(1)

Key Documents

P63395

Sigma-Aldrich

2,4-Pyridinedicarboxylic acid monohydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO4 · H2O
CAS Number:
Molecular Weight:
185.13
Beilstein:
131631
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

246-248 °C (dec.) (lit.)

SMILES string

[H]O[H].OC(=O)c1ccnc(c1)C(O)=O

InChI

1S/C7H5NO4.H2O/c9-6(10)4-1-2-8-5(3-4)7(11)12;/h1-3H,(H,9,10)(H,11,12);1H2

InChI key

PLNFTNOXJDBMSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adeline Sham et al.
Tissue engineering. Part A, 21(5-6), 1106-1115 (2014-11-06)
Clinical applications of tissue engineering are constrained by the ability of the implanted construct to invoke vascularization in adequate extent and velocity. To overcome the current limitations presented by local delivery of single angiogenic factors, we explored the incorporation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service