Skip to Content
Merck
All Photos(2)

Key Documents

P60205

Sigma-Aldrich

2-Pyridinealdoxime methiodide

99%

Synonym(s):

2-PAM, 2-PAM iodide, Pralidoxime iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11IN2O
CAS Number:
Molecular Weight:
266.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

220 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[I-].C[n+]1ccccc1\C=N\O

InChI

1S/C7H8N2O.HI/c1-9-5-3-2-4-7(9)6-8-10;/h2-6H,1H3;1H

InChI key

QNBVYCDYFJUNLO-UHFFFAOYSA-N

Gene Information

human ... ACHE(43)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eduardo A González et al.
Archives of toxicology, 94(6), 2149-2162 (2020-04-19)
Organophosphate (OP) threat agents can trigger seizures that progress to status epilepticus, resulting in persistent neuropathology and cognitive deficits in humans and preclinical models. However, it remains unclear whether patients who do not show overt seizure behavior develop neurological consequences.
Qiqi Zheng et al.
Analytica chimica acta, 982, 78-83 (2017-07-25)
An electrochemical method based on non-enzymatic inhibition for the determination of organophosphorus pesticide (OPPs) using pralidoxime chloride (PAM-Cl) as a universal electrochemical probe was reported. Cyclic voltammetry was performed to characterize the redox properties of pralidoxime and OPPs. Differential pulse
Kimberly D Spradling et al.
Journal of neuroinflammation, 8, 84-84 (2011-07-23)
Although the acute toxicity of organophosphorus nerve agents is known to result from acetylcholinesterase inhibition, the molecular mechanisms involved in the development of neuropathology following nerve agent-induced seizure are not well understood. To help determine these pathways, we previously used
Lorise C Gahring et al.
American journal of physiology. Lung cellular and molecular physiology, 319(4), L683-L692 (2020-07-30)
Nicotine of unprecedented concentrations and purity is being inhaled by those using commercially available electronic nicotine delivery systems (ENDS). The consequences of this route of self-administration on the immunological response to inhaled allergens are not known. In mice, sensitization and
Kevin G McGarry et al.
Toxicological sciences : an official journal of the Society of Toxicology, 174(1), 124-132 (2019-12-28)
Organophosphorus (OP) compounds, which include insecticides and chemical warfare nerve agents (CWNAs) such as sarin (GB) and VX, continue to be a global threat to both civilian and military populations. It is widely accepted that cholinesterase inhibition is the primary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service