Skip to Content
Merck
All Photos(4)

Key Documents

P3515

Sigma-Aldrich

Poly(ethylene glycol)

average Mn 950-1,050

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.07

form

waxy solid

Quality Level

mol wt

average Mn 950-1,050

viscosity

17.4 cSt(210 °F)(lit.)

mp

39 °C (lit.)

solubility

water: soluble (PEG is soluble in water approximately 800 mg/ml, 20 °C)

density

1.0926 g/mL at 60 °C
1.0765 g/mL at 80 °C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) is a non-toxic and water soluble polymer, which can have the tendency to resist recognition by the immune system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Poly (L-lysine)-g-poly (ethylene glycol) layers on metal oxide surfaces: attachment mechanism and effects of polymer architecture on resistance to protein adsorption"
Kenausis.LG, et al.
The Journal of Physical Chemistry B, 104(14), 3298-3309 (2000)
"Biodegradable poly (e-caprolactone)-poly (ethylene glycol) block copolymers: characterization and their use as drug carriers for a controlled delivery system"
Zhou S, et al.
Biomaterials, 24(20), 3563-3570 (2003)
"Poly (ethylene glycol)-containing hydrogels in drug delivery"
Peppas.AN, et al.
J. Controlled Release, 62(1), 81-87 (1999)
"Methoxy poly (ethylene glycol)-poly (lactide)(MPEG-PLA) nanoparticles for controlled delivery of anticancer drugs"
Dong Y and Feng S-S
Biomaterials, 25(14), 2843-2849 (2004)
Yue Hugh Guan et al.
Journal of chromatography. A, 1424, 102-110 (2015-03-31)
Manufacturing high-value added biotech biopharmaceutical products (e.g. therapeutic proteins) requires quick-to-develop, GMP-compliant, easy-to-scale and cost effective preparatory chromatography technologies. In this work, we describe the construction and testing of a set of 5-mm inner diameter stainless steel toroidal columns for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service