All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C20H14
CAS Number:
Molecular Weight:
254.33
Beilstein:
1910570
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
powder
bp
417 °C (lit.)
mp
153-155 °C (lit.)
SMILES string
c1ccc(cc1)-c2c3ccccc3cc4ccccc24
InChI
1S/C20H14/c1-2-8-15(9-3-1)20-18-12-6-4-10-16(18)14-17-11-5-7-13-19(17)20/h1-14H
InChI key
LUBXLGUQZVKOFP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tarun Sharma et al.
European journal of drug metabolism and pharmacokinetics, 45(2), 243-255 (2019-12-02)
Boswellic acids (BAs) include β-boswellic acid (BA), 3-acetyl-β-boswellic acid, 11-keto-β-boswellic acid, 3-acetyl-11-keto-β-boswellic acid, β-boswellic alcohol, and 3-acetyl-11-hydroxy-β-BA from Boswellia species, and are the main active ingredients of Boswellia serrata extracts (BSE). BSE have been used for the treatment of different
John Prakash et al.
Scientific reports, 10(1), 8921-8921 (2020-06-04)
Qualitative and quantitative display of multiple fluorescent analytes is made simple and reliable in this micelle assisted methodology. The adopted method involves micelle assisted evincing of ppb level of PAHs in water; measurement of total fluorescence (white light excitation fluorescence
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service