All Photos(2)
About This Item
Linear Formula:
C6Br5OH
CAS Number:
Molecular Weight:
488.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
96%
form
powder
mp
223-226 °C (lit.)
SMILES string
Oc1c(Br)c(Br)c(Br)c(Br)c1Br
InChI
1S/C6HBr5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H
InChI key
SVHOVVJFOWGYJO-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jana Skopalová et al.
Analytica chimica acta, 999, 60-68 (2017-12-20)
A carbon fiber brush electrode (CFBE) was newly designed and used as a substrate for both controlled potential electrolysis and atmospheric solids analysis probe (ASAP) mass spectrometry. Electropolymerized and strongly adsorbed products of electrolysis were directly desorbed and ionized from
Monika Jarosiewicz et al.
International journal of molecular sciences, 21(11) (2020-06-04)
Brominated flame retardants (BFRs) have been using to reduce the flammability of plastics contained in many products, such as household articles, furniture, mattresses, textiles or insulation. Considering the fact that these compounds may be released into the environment leading to
M Ghosh et al.
Acta crystallographica. Section D, Biological crystallography, 56(Pt 9), 1085-1095 (2000-08-25)
The binding of two organohalogen substances, pentabromophenol (PBP) and 2,4,6-tribromophenol (TBP), to human transthyretin (TTR), a thyroid hormone transport protein, has been studied by in vitro competitive binding assays and by X-ray crystallography. Both compounds bind to TTR with high
J A Szymańska et al.
International journal of occupational medicine and environmental health, 8(3), 245-254 (1995-01-01)
Pentabromophenol (PBP) was administered in a single or repeated doses to mice or in a single dose to rats. Slight changes were noted in the level of SGPT in mice serum and GSH in the liver, with a more pronounced
Monika Jarosiewicz et al.
Chemosphere, 227, 93-99 (2019-04-16)
Currently, more and more concerns are related to oxidative stress appearing in cells as a result of xenobiotics action. It has been found that selected brominated flame retardants (BFRs) can cause reactive oxygen species (ROS) induction at environmental concentrations. Excessive
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service