Skip to Content
Merck
All Photos(1)

Key Documents

OTV000264

Sigma-Aldrich

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2
CAS Number:
Molecular Weight:
193.24
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:

form

solid

InChI

1S/C11H15NO2/c1-13-10-5-8-3-4-12-7-9(8)6-11(10)14-2/h5-6,12H,3-4,7H2,1-2H3

InChI key

CEIXWJHURKEBMQ-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of Otava

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiabing Li et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(24), 7724-7730 (2013-05-17)
The lithiation of N-tert-butoxycarbonyl (N-Boc)-1,2,3,4-tetrahydroisoquinoline was optimized by in situ IR (ReactIR) spectroscopy. Optimum conditions were found by using n-butyllithium in THF at -50 °C for less than 5 min. The intermediate organolithium was quenched with electrophiles to give 1-substituted 1,2,3,4-tetrahydroisoquinolines. Monitoring
The effect of an isoquinoline analogue on voluntary ethanol ingestion in rats.
S Naeger et al.
Proceedings of the Western Pharmacology Society, 34, 35-37 (1991-01-01)
Ken-ichi Umehara et al.
Drug metabolism and disposition: the biological fate of chemicals, 37(7), 1427-1433 (2009-04-11)
(-)-N-{2-[(R)-3-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-2-carbonyl)piperidino]ethyl}-4-fluorobenzamide (YM758), a novel "funny" If current channel (If channel) inhibitor, is developed as a treatment for stable angina and atrial fibrillation. In this study, the pharmacokinetic/pharmacodynamic (PK/PD) relationship after intravenous administration of YM758 to tachycardia-induced dogs was investigated and
S A Barker et al.
Biochemical pharmacology, 30(17), 2461-2468 (1981-09-01)
Gas chromatographic/mass spectrometric data are presented which demonstrate the presence of 6,7-dihydroxy-1,2,3,4-tetrahydroisoquinoline (DHTIQ) as a normal constituent of rat brain. The level of DHTIQ was calculated to be 10.0 +/- 3.0 ng/g wet weight (+/- S.D., N = 9) of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service