OCNQCK525
EDC Conjugation Kit for Carboxylic Cd-based Core/Shell Quantum Dots
fluorescence λem 525 nm, for 4 conjugation
About This Item
fluorescence
λem 525 nm
Looking for similar products? Visit Product Comparison Guide
General description
- QSH
- EDC
- Coupling Buffer
- Quenching Buffer
- Storage Buffer
- Tubes 4 x 1.7 mL
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral
Target Organs
Stomach,large intestine,lymph node
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service