NCXGSGN980
Gold Nanoshells
λmax, 980 nm, NanoXact, 0.05 mg/mL in water, mPEG 5 kDa (Polymer)
About This Item
Recommended Products
form
nanoshells
concentration
0.05 mg/mL in water
λmax
980 nm
Looking for similar products? Visit Product Comparison Guide
Related Categories
1 of 4
This Item | NCXGSPN800 | NCXGSLH800 | NCXGSGH660 |
---|---|---|---|
0.05 mg/mL in water | 0.05 mg/mL in water | 1 mg/mL in water | 1 mg/mL in water |
980 nm | 800 nm | 800 nm | 660 nm |
Application
Our gold nanoshells are surface plasmon resonant (SPR) nanoparticles consisting of a nanoscale silica core surrounded by an ultra-thin gold shell. By changing the ratio of the core diameter and the shell thickness the absorption and scattering properties of the nanoshells can be tuned across the visible and near-IR (NIR) regions. For these shells the increase in the size of the silica core, and decrease in the thickness of the gold shell causes the plasmon resonance to shift toward the NIR region. This class of plasmonic nanoparticles has a wide variety of applications, including uses in optical filters, sensing and cancer therapy. To increase the steric stability our gold nanoshells are coated with PVP (40 kD polyvinylpyrrolidone).
Storage and Stability
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service