NCXAUXR40
Gold Nanospheres
40 nm, for Covalent Conjugation, Bioready, Carboxyl, OD 20, in Water
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
form
nanospheres
OD
20
particle size
40 nm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Gold Nanosphere (40 nm, for covalent conjugation, Bioready, carboxyl, OD 20, in water) can be potentially used in the applications such as drug delivery, photoacoustic imaging, electrochemical immunosensors, and surface plasmon nanopatterning.
Owing to their plasmonic properties nanoparticles of gold and silver find broad range of applications extending from diagnostics to molecular detection, photothermal and microwave-based therapies, and nanotoxicology. Colloidal silver nanoparticles also find applications in antimicrobial coatings, electronics and photovoltaics, and surface enhanced Raman spectroscopy (SERS) whereas the colloidal gold nanoparticles find applications in lateral flow assays (immunochromatographic assays) and as TEM labels and size standards.
Our BioReady materials are engineered for reliable conjugation with antibodies, proteins, peptides, and other biomolecules and are ideal for lateral flow (immunochromatographic assays) and other diagnostics related applications.
The BioReady spherical gold nanoparticles with carboxyl surface can be used for robust covalent conjugation. This allows for greater stability, sensitivity, reproducibility, and control over particle antibody loading (often requires less antibody compared to passive adsorption). Our covalently-bound streptavidin conjugated gold nanoparticles allow easy binding with biotin.
The BioReady conjugation kits contain dried gold nanoparticles with NHS-activated carboxyl surface. With pre-functionalized nanoparticles with EDC/NHS surface, these are well suited for rapid antibody screening.
The small reaction NHS kits contain 50 μL of 20 OD gold in each unit whereas the large reaction kits contain 500 μL of 20 OD gold in each unit.
Our BioReady materials are engineered for reliable conjugation with antibodies, proteins, peptides, and other biomolecules and are ideal for lateral flow (immunochromatographic assays) and other diagnostics related applications.
The BioReady spherical gold nanoparticles with carboxyl surface can be used for robust covalent conjugation. This allows for greater stability, sensitivity, reproducibility, and control over particle antibody loading (often requires less antibody compared to passive adsorption). Our covalently-bound streptavidin conjugated gold nanoparticles allow easy binding with biotin.
The BioReady conjugation kits contain dried gold nanoparticles with NHS-activated carboxyl surface. With pre-functionalized nanoparticles with EDC/NHS surface, these are well suited for rapid antibody screening.
The small reaction NHS kits contain 50 μL of 20 OD gold in each unit whereas the large reaction kits contain 500 μL of 20 OD gold in each unit.
Storage and Stability
DO NOT FREEZE. Store at 2-8C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Legal Information
Product of nanoComposix, Inc.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Surface plasmon-based nanopatterning assisted by gold nanospheres
Heltzel A, et al.
Nanotechnology, 19(2), 025305-025305 (2007)
Photoacoustic imaging of living mouse brain vasculature using hollow gold nanospheres
Lu W, et al.
Biomaterials, 31(9), 2617-2626 (2010)
Ultrasensitive electrochemical immunosensor for clinical immunoassay using thionine-doped magnetic gold nanospheres as labels and horseradish peroxidase as enhancer
Tang D, et al.
Analytical Chemistry, 80(5), 1582-1588 (2008)
Exceptionally high payload of doxorubicin in hollow gold nanospheres for near-infrared light-triggered drug release
You J, et al.
ACS Nano, 4(2), 1033-1041 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service