N16428
4-Nitrocinnamic acid, predominantly trans
97%
Synonym(s):
3-(4-Nitrophenyl)-2-propenoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
O2NC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
193.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
powder
mp
289 °C (dec.) (lit.)
SMILES string
OC(=O)\C=C\c1ccc(cc1)[N+]([O-])=O
InChI
1S/C9H7NO4/c11-9(12)6-3-7-1-4-8(5-2-7)10(13)14/h1-6H,(H,11,12)/b6-3+
InChI key
XMMRNCHTDONGRJ-ZZXKWVIFSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
María L Salum et al.
Journal of photochemistry and photobiology. B, Biology, 148, 128-135 (2015-04-29)
Cinnamic acids are present in all kinds of plant tissues and hence in herbs and derived medicines, cosmetics and foods. The interest in their role in plants and their therapeutic applications has grown exponentially. Because of their molecular structure they
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service