Key Documents
MT59904591
Mettler Toledo RedCap pH electrode easy clean
L 120 mm × 12 mm
About This Item
UNSPSC Code:
41115604
NACRES:
NB.79
material
glass (A41)
glass membrane (polymer)
description
S7 connection
Without cable
manufacturer/tradename
Mettler-Toledo 59904591
L × W
120 mm × 12 mm
L
120 mm × 12 mm
General description
Mettler Toledo RedCap pH electrode 405-60-T-S7/120/9848 is a generalist. It can be used as a puncture electrode in soft media, but also for general aqueous samples. Thanks to the triple ceramic junction and huge glass membrane the 405-60-T-S7/120/9848 responds really quickly. The electrolyte is free from silver due to the ARGENTHAL reference system and the silver ion trap.
  • Combination pH electrode
  • Easy to clean cone-shaped tip
  • Triple ceramic diaphragm
  • Large A41 glass membrane
  • Detachable cable
Legal Information
ARGENTHAL is a trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)