MT30266941
Mettler Toledo FiveEasy™ Plus pH / mV bench meter
model, FP20, TRIS Kit including glass LE420 pH electrode with moveable sleeve diaphragm, AC/DC input 230 V AC, universal plug set
Synonym(s):
Benchtop pH meter, Fiveeasy plus meter, pH meter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115603
NACRES:
NB.79
Recommended Products
feature
BNC/cinch connector
display 4.3 in. LCD
model FP20
pH measuring range
-2-16 pH unit, accuracy: ±0.01, resolution: 0.01
temperature measuring range
-5-105 °C, accuracy: ±0.3 °C, resolution: 0.1 °C
battery power
12 V
Looking for similar products? Visit Product Comparison Guide
General description
Ideally suited for applications in the laboratory, the new FiveEasy and FiveEasy Plus benchtop instruments provide high quality pH/mV or conductivity measurements with the simple click of a button. Compact in design, and featuring various data export opportunities, the FiveEasy series instruments are the perfect companion to get reliable measurement data, all at a reasonable price.
Features and Benefits
Intuitive Operation – Measurement Made Easy
Featuring a large, well-structured display for easy viewing, the instruments′ intuitive button arrangement and simple menu ensure measurements can be performed in just a few clicks.
Compact Design –Save Space
Thanks to its compact design, little bench space is needed for the Five series meters. The sensor holder can be easily removed and stored on the side of the meter after use.
Fast Connectivity – Simple and Secure Data Trasnsfer
The FiveEasy Plus series meters allows data to be exported either directly to a printer or a PC for further processing using its RS232 or USB ports.
Featuring a large, well-structured display for easy viewing, the instruments′ intuitive button arrangement and simple menu ensure measurements can be performed in just a few clicks.
Compact Design –Save Space
Thanks to its compact design, little bench space is needed for the Five series meters. The sensor holder can be easily removed and stored on the side of the meter after use.
Fast Connectivity – Simple and Secure Data Trasnsfer
The FiveEasy Plus series meters allows data to be exported either directly to a printer or a PC for further processing using its RS232 or USB ports.
Other Notes
Ideally suited for demanding, ′dirty′ samples such as emulsions, oily samples, waste water, TRIS, etc.
Legal Information
FiveEasy is a trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service