MP150105
MakroPor, silicon membrane
thickness 50 μm, pore diameter 1 μm, pore size 1.5 μm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Linear Formula:
Si
UNSPSC Code:
12352103
NACRES:
NA.23
Recommended Products
description
FZ single crystal Si
Pore geometry: trigonal arrangement
packaging
pkg of 5 pieces
size
10 mm × 10 mm
thickness
50 μm
pore size
1 μm pore diameter
1.5 μm (interpore distance)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Macro porous silicon is useful for applications requiring mechanical and chemical stability with high order of the pores. Our macro porous silicon membranes are manufactured from single crystal n-doped silicon wafers and possess pore diameter between 1 to 8 μm. The standard deviation of pore diameter and interpore distance for these porous silicon membrane materials is less than 5 %. These macro porous silicon finds applications in filtration, as platforms for multi-functional flow-through sensors, as fuels cells electrodes or micro reactors etc. They can also be used as templates for the fabrication of micro scale composites, such as tubes or wires by electrochemical deposition or by using polymer melts. We suggest using vacuum tweezers on the front side or polymer tweezers on the side faces.
Legal Information
Product of SmartMembranes-GmbH
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service