Recommended Products
vapor pressure
47.6 mmHg ( 37.7 °C)
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.52 (lit.)
bp
113 °C (lit.)
mp
−63 °C (lit.)
density
1.014 g/mL at 25 °C (lit.)
SMILES string
Cc1cccs1
InChI
1S/C5H6S/c1-5-3-2-4-6-5/h2-4H,1H3
InChI key
XQQBUAPQHNYYRS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
59.0 °F - closed cup
Flash Point(C)
15 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chemistry (Weinheim an der Bergstrasse, Germany), 21(32), 11483-11494 (2015-07-02)
To harvest energy from the near-infrared (near-IR) and infrared (IR) regions of the electromagnetic spectrum, which constitutes nearly 70 % of the solar radiation, there is a great demand for near-IR and IR light-absorbing sensitizers that are capable of undergoing ultrafast
Food research international (Ottawa, Ont.), 123, 317-326 (2019-07-10)
Detection thresholds are used routinely to determine the odour-active compounds in foods. The composition of a food matrix, such as hydrophobicity or solids content, has an impact on the release of flavour compounds, and thus on thresholds. In the case
Journal of the American Chemical Society, 141(13), 5350-5358 (2019-03-07)
Electronic structure modulation of metal-organic frameworks (MOFs) through the connection of linker "wires" as a function of an external stimulus is reported for the first time. The established correlation between MOF electronic properties and photoisomerization kinetics as well as changes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service