Recommended Products
material
silica coating (SiO2)
description
die layer thickness 300 nm SiO2
electrode layer thickness 60 nm gold
feature
electrode large serpentine
manufacturer/tradename
LS100
die L × W × thickness
20 mm × 10 mm × 525 μm, silicon
electrode size
10 nm , chrome
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service