HAM61614-01
Fill tubing for Hamilton® Microlab® 600 pipettor
FEP, gauge × L 12 × 1219 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
material
FEP
gauge × L
12 × 1219 mm
tubing volume
4571 μL
fitting
1/4-28 non-tapered
suitability
suitable for (use with greater than 1 mL syringes)
Looking for similar products? Visit Product Comparison Guide
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service