Skip to Content
Merck
All Photos(2)

Key Documents

H18801

Sigma-Aldrich

3′-Hydroxyacetophenone

97%

Synonym(s):

1-(3-Hydroxyphenyl)ethanone, 3-Acetylphenol, 3-Hydroxyphenylethanone, m-Hydroxyacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COCH3
CAS Number:
Molecular Weight:
136.15
Beilstein:
2040676
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

bp

296 °C (lit.)

mp

90-95 °C (lit.)

density

1.1 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1cccc(O)c1

InChI

1S/C8H8O2/c1-6(9)7-3-2-4-8(10)5-7/h2-5,10H,1H3

InChI key

LUJMEECXHPYQOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

247.3 °F - closed cup

Flash Point(C)

119.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Otani et al.
Bioorganic & medicinal chemistry letters, 18(12), 3582-3584 (2008-05-31)
m-Acetylphenyl-beta-d-glucopyranosides and m-acetylphenyl-alpha/beta-d-mannopyranosides were synthesized by the Koenigs-Knorr, Mitsunobu, and Helferich reactions as key glycosylation reactions, respectively. Their spectroscopic properties and antioxidative activities were characterized as potential ultraviolet B-ray absorbents.
Stefania Ferrari et al.
Journal of medicinal chemistry, 54(1), 211-221 (2010-12-04)
Folate analogue inhibitors of Leishmania major pteridine reductase (PTR1) are potential antiparasitic drug candidates for combined therapy with dihydrofolate reductase (DHFR) inhibitors. To identify new molecules with specificity for PTR1, we carried out a virtual screening of the Available Chemicals
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service