GF92376445
Cobalt/Silicon/Boron/Iron/Nickel
foil, thickness 0.025 mm, Co66%/Si15%/B 14%/Fe 4%/Ni 1%, length 5 m, coil width 25 mm
Synonym(s):
CO810250
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
foil
manufacturer/tradename
Goodfellow 923-764-45
L
5 m
coil width
25 mm
thickness
0.025 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service