Skip to Content
Merck
All Photos(2)

Key Documents

GF80154334

Tungsten

wire reel, 25m, diameter 0.15mm, clean, 99.95%

Synonym(s):

Tungsten, W 005145, W

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
EC Number:
MDL number:
UNSPSC Code:
12141747
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

wire

manufacturer/tradename

Goodfellow 801-543-34

resistivity

4.9 μΩ-cm, 20°C

L × diam.

25 m × 0.15 mm

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[W]

InChI

1S/W

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard R Schrock et al.
Angewandte Chemie (International ed. in English), 42(38), 4592-4633 (2003-10-09)
Catalytic olefin metathesis has quickly emerged as one of the most often-used transformations in modern chemical synthesis. One class of catalysts that has led the way to this significant development are the high-oxidation-state alkylidene complexes of molybdenum. In this review
Synthetic analogues and reaction systems relevant to the molybdenum and tungsten oxotransferases.
John H Enemark et al.
Chemical reviews, 104(2), 1175-1200 (2004-02-12)
Elias Papaconstantinou et al.
Frontiers in bioscience : a journal and virtual library, 8, s813-s825 (2003-09-06)
Polyoxometalates (POM) have a rich and promising photochemical behavior. Upon illumination at the O M CT band (i.e., in UV and near-visible area) become powerful oxidizing reagents able to oxidize various organic compounds. In the process POM undergo stepwise reduction
Mehran Ahmadlou et al.
Nature communications, 6, 6773-6773 (2015-04-04)
The superior colliculus is a layered structure important for body- and gaze-orienting responses. Its superficial layer is, next to the lateral geniculate nucleus, the second major target of retinal ganglion axons and is retinotopically organized. Here we show that in
Nikolay Strigul
Ecotoxicology and environmental safety, 73(6), 1099-1113 (2010-09-16)
Tungsten is a widely used transition metal that has not been thoroughly investigated with regards to its ecotoxicological effects. Tungsten anions polymerize in environmental systems as well as under physiological conditions in living organisms. These polymerization/condensation reactions result in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service