Skip to Content
Merck
All Photos(2)

Key Documents

GF69773181

Titanium

wire reel, 100m, diameter 1.0mm, as drawn, 99.6+%

Synonym(s):

Titanium, TI005129

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23

Assay

99.6%

form

wire

autoignition temp.

860 °F

manufacturer/tradename

Goodfellow 697-731-81

resistivity

42.0 μΩ-cm, 20°C

L × diam.

100 m × 1.0 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Qiang Yu et al.
Journal of nanoscience and nanotechnology, 14(6), 4387-4393 (2014-04-18)
The TiO2 nanotubes by anodization have been extensively studied for medical implant and orthopedic applications because of enhancing bone development. In the present study, a new nano-foveolae structure verified by SEM and AFM was prepared by simulating the nanotubes exfoliation
Shaoming Yang et al.
Journal of nanoscience and nanotechnology, 14(6), 4292-4296 (2014-04-18)
The structural and functional characterizations of titanium nitride (TiN) advanced ceramic microsprings (CMSs), with a coil diameter of several micrometers and synthesized by chemical vapor deposition (CVD) were investigated by microscopy techniques. The CMSs were sufficiently mechanically elastic for extension
Bijuan Zheng et al.
Journal of nanoscience and nanotechnology, 14(5), 3527-3531 (2014-04-17)
Pd-modified N-doped TiO2 nanoparticles were prepared by the sol-gel method. The X-ray diffraction (XRD) pattern indicated that the pure anatase TiO2 has been obtained. Transmission electron microscope (TEM) was used to observe the micro-morphology of the nanoparticles. The average size
Yupeng Zhang et al.
Journal of nanoscience and nanotechnology, 14(6), 4303-4307 (2014-04-18)
Na2Ti3O7 nanotubes was synthesized by a hydrothermal method, and characterized by X-ray diffraction (XRD), scanning electron microscopy (SEM), and transmission electron microscopy (TEM). Then the material was coated on Al2O3 ceramic substrate to fabricate humidity sensors using Ag-Pd as interdigitated
Dongsheng Guan et al.
Journal of nanoscience and nanotechnology, 14(6), 4541-4550 (2014-04-18)
Bamboo-type TiO2 nanotube arrays with high surface area can be synthesized by alternating voltage (AV) anodization for their important use as photocatalytic medium. Their morphologies are highly dependent on preparation parameters including anodization time and electrolyte composition. Minimum time of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service