Skip to Content
Merck
All Photos(2)

Key Documents

GF64762140

Molybdenum

foil, 50mm disks, thickness 0.25mm, annealed, 99.9%

Synonym(s):

Molybdenum, MO000370

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9%

form

foil

manufacturer/tradename

Goodfellow 647-621-40

resistivity

5.0 μΩ-cm, 20°C

size × thickness

50 mm × 0.25 mm

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles F Baker et al.
Dalton transactions (Cambridge, England : 2003), (9)(9), 1298-1303 (2004-07-15)
New molybdenum(VI) nitride oxides were synthesised by the reaction of strontium nitride and calcium nitride with molybdenum foil at high temperature in sealed stainless steel crucibles. The reactions yielded single crystalline products determined by X-ray diffraction to form complex structures
J G Watkins et al.
The Review of scientific instruments, 79(10), 10F125-10F125 (2008-12-03)
Two modular arrays of Langmuir probes designed to handle a heat flux of up to 25 MW/m(2) for 10 s exposures have been installed in the lower divertor target plates of the DIII-D tokamak. The 20 pyrolytic graphite probe tips
Dattatray J Late et al.
Nanotechnology, 19(26), 265605-265605 (2008-07-02)
Nanocrystalline lanthanum hexaboride (LaB(6)) films have been deposited on molybdenum foil by the pulsed laser deposition (PLD) technique. The as-deposited films were characterized by x-ray diffraction (XRD), scanning electron microscopy (SEM) and atomic force microscopy (AFM). The XRD pattern shows
Lidia Regoli et al.
The Science of the total environment, 435-436, 96-106 (2012-08-01)
In a regulatory context, bioaccumulation or bioconcentration factors are used for considering secondary poisoning potential and assessing risks to human health via the food chain. In this paper, literature data on the bioaccumulation of molybdenum in the aquatic organisms are
Tingyu Shi et al.
Journal of cellular biochemistry, 112(10), 2721-2728 (2011-06-17)
When intracelluar pathogens enter the host macrophages where in addition to oxidative and antibiotic mechanisms of antimicrobial activity, nutrients are deprived. Human pathogen Mycobacterium tuberculosis is one of macrophage parasitisms, which can replicate and persist for decades in dormancy state

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service