GF59666945
Chromium
rod, 10mm, diameter 5.0mm, 99.7+%
Synonym(s):
Chromium, CR007920
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
≥99.7%
form
rod
manufacturer/tradename
Goodfellow 596-669-45
resistivity
12.7 μΩ-cm, 20°C
L × diam.
10 mm × 5.0 mm
bp
2672 °C (lit.)
mp
1857 °C (lit.)
density
7.14 g/mL at 25 °C (lit.)
SMILES string
[Cr]
InChI
1S/Cr
InChI key
VYZAMTAEIAYCRO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Reviews of environmental contamination and toxicology, 217, 75-140 (2012-02-22)
Bioremediation offers the possibility of using living organisms (bacteria, fungi, algae,or plants), but primarily microorganisms, to degrade or remove environmental contaminants, and transform them into nontoxic or less-toxic forms. The major advantages of bioremediation over conventional physicochemical and biological treatment
Critical reviews in toxicology, 43(3), 244-274 (2013-03-01)
Abstract Chronic exposure to high concentrations of hexavalent chromium (Cr(VI)) in drinking water causes intestinal adenomas and carcinomas in mice, but not in rats. Cr(VI) causes damage to intestinal villi and crypt hyperplasia in mice after only one week of
The Journal of nutritional biochemistry, 23(4), 313-319 (2012-03-20)
Type 2 diabetes is often associated with obesity, dyslipidemia and cardiovascular anomalies and is a major health problem approaching global epidemic proportions. Insulin resistance, a prediabetic condition, precedes the onset of frank type 2 diabetes and offers potential avenues for
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(22), 5733-5744 (2014-09-13)
Anaplastic thyroid carcinoma (ATC) is one of the most aggressive forms of cancer with no curative therapies available. To date, strategies to target ATC by immunotherapy have not been evaluated. We investigated whether ATC would be a suitable target for
Environment international, 72, 176-185 (2014-05-06)
Rapid development, industrialisation, and urbanisation have resulted in serious contamination of soil by metals and metalloids from anthropogenic sources in many areas of the world, either directly or indirectly. Exponential urban and economic development has resulted in human populations settling
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service