Skip to Content
Merck
All Photos(2)

Key Documents

GF51626831

Molybdenum

rod, 100mm, diameter 2.0mm, centerless ground, 99.9%

Synonym(s):

Molybdenum, MO007903

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
MDL number:
UNSPSC Code:
12141727
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

rod

manufacturer/tradename

Goodfellow 516-268-31

resistivity

5.0 μΩ-cm, 20°C

L × diam.

100 mm × 2.0 mm

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernd Masepohl et al.
Advances in experimental medicine and biology, 675, 49-70 (2010-06-10)
The vast majority of the purple nonsulfur photosynthetic bacteria are diazotrophs, but the details of the complex regulation of the nitrogen fixation process are well understood only for a few species. Here we review what is known of the well-studied
Lidia Regoli et al.
The Science of the total environment, 435-436, 96-106 (2012-08-01)
In a regulatory context, bioaccumulation or bioconcentration factors are used for considering secondary poisoning potential and assessing risks to human health via the food chain. In this paper, literature data on the bioaccumulation of molybdenum in the aquatic organisms are
Tingyu Shi et al.
Journal of cellular biochemistry, 112(10), 2721-2728 (2011-06-17)
When intracelluar pathogens enter the host macrophages where in addition to oxidative and antibiotic mechanisms of antimicrobial activity, nutrients are deprived. Human pathogen Mycobacterium tuberculosis is one of macrophage parasitisms, which can replicate and persist for decades in dormancy state
Guenter Schwarz et al.
Metal ions in life sciences, 13, 415-450 (2014-01-29)
Molybdenum is an essential trace element and crucial for the survival of animals. Four mammalian Mo-dependent enzymes are known, all of them harboring a pterin-based molybdenum cofactor (Moco) in their active site. In these enzymes, molybdenum catalyzes oxygen transfer reactions
Hanmei Hu et al.
Journal of nanoscience and nanotechnology, 14(6), 4462-4468 (2014-04-18)
A mild one-pot hydrothermal route has been successfully designed to controllably prepare orthorhombic alpha-MoO3 nanobelts and monoclinic MoO2 microaxletrees respectively by adjusting the dosage of (NH4)6M07O24 x 4H2O (AHM). The products are characterized by X-ray diffraction (XRD), field-emission scanning electron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service