GF39275208
Polytetrafluoroethylene
(PTFE), film, thickness 0.1 mm, L 10 m, coil width 610 mm
Synonym(s):
FP301301
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
film
manufacturer/tradename
Goodfellow 392-752-08
L
10 m
coil width
610 mm
thickness
0.1 mm
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service