GF38854187
Nickel-chromium alloy, Ni80Cr20
5m insulated wire, 0.025mm conductor diameter, 0.004mm insulation thickness, epoxy insulation
Synonym(s):
NI055850
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
Recommended Products
description
epoxy insulation
form
wire (insulated)
manufacturer/tradename
Goodfellow 388-541-87
L
5 m
conductor diam.
0.025 mm
insulation thickness
0.004 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service