GF37585378
Aluminum
foam, thickness 30 mm, size 100 × 100 mm, bulk density 0.2 g/cm3
Synonym(s):
AL003861
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
autoignition temp.
1400 °F
manufacturer/tradename
Goodfellow 375-853-78
resistivity
2.6548 μΩ-cm
size
100 × 100 mm
thickness
30 mm
bp
2460 °C (lit.)
mp
660.37 °C (lit.)
density
2.7 g/mL at 25 °C (lit.)
bulk density
0.2 g/cm3
SMILES string
[Al]
InChI
1S/Al
InChI key
XAGFODPZIPBFFR-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service