Skip to Content
Merck
All Photos(2)

Key Documents

GF26536057

Tantalum

foil, not light tested, 25x25mm, thickness 0.004mm, 99.9%

Synonym(s):

Tantalum, TA000130, Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

99.90%

form

foil

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 265-360-57

resistivity

13.5 μΩ-cm, 20°C

size × thickness

25x25 mm × 0.004 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation of the brachial segments of the spinal cord of the chick embryo by means of tantalum foil blocks.
V HAMBURGER
The Journal of experimental zoology, 103(1), 113-142 (1946-10-01)
Xinjian Feng et al.
Nano letters, 10(3), 948-952 (2010-02-12)
Tantalum nitride (Ta3N5) has a band gap of approximately 2.07 eV, suitable for collecting more than 45% of the incident solar spectrum energy. We describe a simple method for scale fabrication of highly oriented Ta3N5 nanotube array films, by anodization
H H Pieper et al.
The Review of scientific instruments, 83(5), 055110-055110 (2012-06-07)
We present a versatile system for the preparation of oxide crystal surfaces in the ultra-high vacuum (UHV) at temperatures up to 1300 K. Thermal treatment is accomplished by direct current heating of a tantalum foil in contact with the oxide
Zhongli Cai et al.
Radiation research, 165(3), 365-371 (2006-02-24)
Both thick and thin films of pGEMR-3Zf- plasmid DNA deposited on a tantalum foil were exposed to soft X rays (effective energy of 14.8 keV) for various times in air under a relative humidity of 45% (Gamma approximately 6, where
Arthroplasty of the temporomandibular joint in children with interposition of tantalum foil.
G W N EGGERS
The Journal of bone and joint surgery. American volume, 28, 603-606 (1946-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service