Skip to Content
Merck
All Photos(2)

Key Documents

GF17608529

Tungsten

foil, not light tested, 25x25mm, thickness 0.003mm, 99.95%

Synonym(s):

Tungsten, W 000090, W

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
MDL number:
UNSPSC Code:
12141747
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 176-085-29

resistivity

4.9 μΩ-cm, 20°C

size × thickness

25 x 25 mm × 0.003 mm

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[W]

InChI

1S/W

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Marbach et al.
Physics in medicine and biology, 26(3), 435-443 (1981-05-01)
Curves relating beam energy, scattering foil thickness, central-axis depth-dose, and beam flatness have been generated using data taken on a Siemens 200A betatron. The curve set allows a single combination of tungsten foil thickness and electron beam energy to be
Congshang Wan et al.
Ultramicroscopy, 119, 106-110 (2011-11-15)
Thermionic electron emission from 200 to 500 nm thick coatings of scandium oxide on tungsten foil have been examined in thermionic emission microscopy, spectroscopic photoelectron microcopy, synchrotron radiation and ultraviolet photoelectron spectroscopy (UPS). A clear dependence of the scandium oxide-W
Mehran Ahmadlou et al.
Nature communications, 6, 6773-6773 (2015-04-04)
The superior colliculus is a layered structure important for body- and gaze-orienting responses. Its superficial layer is, next to the lateral geniculate nucleus, the second major target of retinal ganglion axons and is retinotopically organized. Here we show that in
P Thongkumkoon et al.
Toxicology letters, 226(1), 90-97 (2014-02-08)
The toxicity of nanomaterials has been well known, but mechanisms involved have been little known. This study was aimed at looking at direct interaction between nanomaterials and naked DNA for some fundamental understanding. Two different types of nanomaterials, carbon nanotubes
Jan R Andreesen et al.
Annals of the New York Academy of Sciences, 1125, 215-229 (2007-12-22)
The history and changing function of tungsten as the heaviest element in biological systems is given. It starts from an inhibitory element/anion, especially for the iron molybdenum-cofactor (FeMoCo)-containing enzyme nitrogenase involved in dinitrogen fixation, as well as for the many

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service