Skip to Content
Merck
All Photos(2)

Key Documents

GF10151873

Tantalum

foil, 50x50mm, thickness 1.0mm, annealed, 99.9%

Synonym(s):

Tantalum, TA000490, Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

≥99.9%

form

foil

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 101-518-73

resistivity

13.5 μΩ-cm, 20°C

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingxun Du et al.
Journal of hazardous materials, 225-226, 21-27 (2012-05-23)
In order to overcome the limitation of the application of nanoparticles, tantalum (oxy)nitrides nanotube arrays on a Ta foil were synthesized and introduced in vis (visible light)-Fenton-like system to enhance the degradation of atrazine. At first, the anodization of tantalum
Radmila Panajotovic et al.
Radiation research, 165(4), 452-459 (2006-04-04)
We determined effective cross sections for production of single-strand breaks (SSBs) in plasmid DNA [pGEM 3Zf(-)] by electrons of 10 eV and energies between 0.1 and 4.7 eV. After purification and lyophilization on a chemically clean tantalum foil, dry plasmid
Tantalum foil cuffs in peripheral nerve surgery.
E E CLIFFTON
Surgery, 23(3), 507-514 (1948-03-01)
T Y Hung et al.
Applied optics, 31(22), 4397-4404 (1992-08-01)
A simple four-element fused-silica lens is presented that has a focal length of 31.2 mm and a relative aperture of f/1 for use as a focusing lens for deep UV laser processing. The curvature of the lens is designed with
Tantalum foil pegs.
E MAXWELL
American journal of ophthalmology, 34(3), 444-444 (1951-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service