Skip to Content
Merck
All Photos(2)

Key Documents

GF07560435

Lead

rod, 100mm, diameter 6.35mm, 99.95%

Synonym(s):

Lead, PB007920

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.95%

form

rod

manufacturer/tradename

Goodfellow 075-604-35

resistivity

20.65 μΩ-cm

L × diam.

100 mm × 6.35 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cognitive declines after deep brain stimulation are likely to be attributable to more than caudate penetration and lead location.
Takashi Morishita et al.
Brain : a journal of neurology, 137(Pt 5), e274-e274 (2014-02-13)
Min-Yi Huang et al.
Ecotoxicology and environmental safety, 104, 310-316 (2014-04-15)
Lead (Pb) is a common heavy metal in the natural environment, but its concentration has been increasing alongside widespread industrial and agricultural development in China. The dark-spotted frog Pelophylax (formerly Rana) nigromaculata (Anura: Ranidae) is distributed across East Asia and
Reply: Cognitive declines after deep brain stimulation are likely to be attributable to more than caudate penetration and lead location.
Karsten Witt et al.
Brain : a journal of neurology, 137(Pt 5), e275-e275 (2014-02-20)
Paraskevi Malea et al.
Ecotoxicology and environmental safety, 104, 175-181 (2014-04-01)
The effects of lead uptake on microtubule integrity and cell viability in intermediate-juvenile leaf blades of the seagrass Cymodocea nodosa were investigated under laboratory conditions in increasing exposure concentrations (0.1, 0.25, 0.5, 5, 10, 20 and 40mg/L). Uptake kinetics was
Yunbiao Wang et al.
Ecotoxicology and environmental safety, 104, 317-322 (2014-04-15)
In the nematode Caenorhabditis elegans, stress resistance can be regulated by dauer formation (daf) genes. In the present study, regulation of heavy metal lead (Pb) toxicity by the 90-kDa heat shock proteins (Hsp90; daf-21) was investigated in both wild-type C.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service