Key Documents
ESYNTH007
Sigma-Aldrich
SynLectro PTFE Electrode Holder
About This Item
UNSPSC Code:
81141502
NACRES:
NB.61
Pricing and availability is not currently available.
General description
SynLectro PTFE Electrode Holder is part of the SynLectro Electrolysis Platform for synthetic electrochemistry. This electrode holder is designed to hold electrodes in firmly place to create a homogenous electric field. The PTFE holder prevents damage and scratching to the electrode surface - for use with soft electrode materials.
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Related Content
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.