DWK986702
WHEATON® liquid scintillation vial with attached foil lined urea cap, HDPE
lips on vial transparent high-density polyethylene bottle, capacity (20 mL), screw cap
Synonym(s):
WHEATON® liquid scintillation vial with attached foil lined urea cap, glass, 20mL, HDPE vials, Liquid scintillation vials, Scintillation, WHEATON vials, caps attached, metal foil cork liner, vials
About This Item
Recommended Products
material
lips on vial transparent high-density polyethylene bottle
metal foil liner
white cap
feature
with cap
packaging
pkg of 500 ea
manufacturer/tradename
WHEATON® 986702
bottle diam. × H × W
(27 mm) (61 mm) (302 mm)
capacity
(20 mL)
fitting
22-400 neck
22-400
Related Categories
Features and Benefits
- Converted from Type I borosilicate glass tubing, PET, or HDPE
- Background counts are consistent and low; ultraviolet transmission is high
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)