DWK90K16125F
KIMBLE® disposable screw thread culture tube
flat bottom borosilicate glass, tube diam. × L (16 mm) (125 mm), non-sterile
Synonym(s):
90K16125F
About This Item
material
flat bottom borosilicate glass
flat bottom
screw top neck
transparent borosilicate glass tube
sterility
non-sterile
feature
without cap
packaging
case of 1000 ea Modular tray shrink-wrapped in plastic film
manufacturer/tradename
Kimble® 90K16125F
tube diam. × L
(16 mm) (125 mm)
fitting
neck (External Thread)
General description
- These tubes are suitable for tissue culture work and general bacteriological use.
- Modular trays are shrink-wrapped in plastic film
- Supplied without caps
- No marking spot
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)