DWK73808-15415
Kimble® Closed Top Polypropylene Screw Thread Cap With Welded Ptfe-Faced Silicone Liner
15-415
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.17
Recommended Products
material
PTFE/silicone liner
red polypropylene screw cap (Push fit)
packaging
pkg of 1 packs
manufacturer/tradename
Kimble® 73808-15415
fitting
thread
Related Categories
Application
Closure solution for vials and bottles
Features and Benefits
- PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
- Closures are ideal for repeated autoclaving
- Welded liner technology eliminates the possibility of glue contamination
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)