DWK73750-16150
KIMBLE® disposable screw thread culture tube with white marking spot
round bottom borosilicate glass, tube diam. × L (16 mm) (105 mm), non-sterile
About This Item
material
round bottom borosilicate glass
round bottom
screw top neck
transparent borosilicate glass tube
sterility
non-sterile
feature
without cap
packaging
case of 1000 ea Modular tray shrink-wrapped in plastic film
manufacturer/tradename
Kimble® 73750-16150
tube diam. × L
(16 mm) (105 mm)
tube diam.
(16 mm)
fitting
neck (External Thread)
General description
- These culture tubes are suitable for tissue culture work and general bacteriological use.
- Round-bottomed
- Each tube has a chevron marking area
- Supplied without caps
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)