Skip to Content
Merck
All Photos(1)

Key Documents

DWK258523107

DURAN® Filter funnel

glass, flask volume (95 mL), porosity 1

Synonym(s):

258523107

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104925
NACRES:
NB.43
Pricing and availability is not currently available.

material

glass

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 258523107

disc diam.

95 mm

flask volume

(95 mL)

pore size

1 porosity

capacity

500 mL capacity

Looking for similar products? Visit Product Comparison Guide

General description

One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Büchner funnels are cylindrical with an integral sintered disc. Without additional filter paper, only coarse filtration is possible. Büchner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.

Features and Benefits

• Manufactured from 3.3. expansion borosilicate glass• Funnels have an integral sintered glass filtration disc• Comprehensive range of funnel sizes and sinter porosities (grades) are available• The use of filter paper is not required• Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid• Filter funnels can be used with a filtering flask via a conical rubber seal (GUKO)• Fired-on, highly durable white print

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service