DWK241253708
Duran® Dropping Funnel
pressure tubing with pressure equalisation tube
, standard ground cone 29/32, stopcock and key retaining device, capacity 250 mL
Synonym(s):
241253708
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 241253708
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
Application
uniform and metered liquid supply to a reaction mixture. The rate of supply can be adjusted
Features and Benefits
With pressure equalisation tube, standard ground cone, standard ground stopcock and retaining device; cylindrical, with scale, ground joint and pressure equalisation tube
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service