DWK231752352
Disposable Culture Tube
Synonym(s):
231752352
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121706
NACRES:
NB.13
Recommended Products
material
polypropylene screw cap
soda-lime glass vial
Joint
Joints (GL 18)
feature
seal (rubber)
packaging
pkg of 100 ea
manufacturer/tradename
231752352
W × L
18 mm × 180 mm
Looking for similar products? Visit Product Comparison Guide
Application
Preparation cell culture, sample collecting, storage
Features and Benefits
straight rim, round bottom, not reusable, with screw cap
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service