DWK11-0055N
Wheaton® Microliter Snap Caps With Septa
natural PTFE/silicone/PTFE cap
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.11
Recommended Products
material
natural PTFE/silicone/PTFE cap
packaging
pkg of 100 ea
manufacturer/tradename
WHEATON® 11-0055N
Related Categories
Application
Chromatography
Features and Benefits
- 11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.
- Where noted, the Ring is installed between cap and septa for robotics employing magnetics to move vials.
- Approved for GC and LC (where noted).
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.