Skip to Content
Merck
All Photos(1)

Key Documents

D40809

Sigma-Aldrich

1,2-Dibromoethylene, mixture of cis and trans

98%

Synonym(s):

Acetylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH=CHBr
CAS Number:
Molecular Weight:
185.85
Beilstein:
1719351
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.5405 (lit.)

bp

110 °C/754 mmHg (lit.)

density

2.246 g/mL at 25 °C (lit.)

SMILES string

Br\C=C\Br

InChI

1S/C2H2Br2/c3-1-2-4/h1-2H/b2-1+

InChI key

UWTUEMKLYAGTNQ-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service