D36003
3,4-Dibenzyloxybenzaldehyde
98%
Synonym(s):
3,4-Bis(benzyloxy)benzaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C6H5CH2O)2C6H3CHO
CAS Number:
Molecular Weight:
318.37
Beilstein:
1132811
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
91-94 °C (dec.) (lit.)
SMILES string
[H]C(=O)c1ccc(OCc2ccccc2)c(OCc3ccccc3)c1
InChI
1S/C21H18O3/c22-14-19-11-12-20(23-15-17-7-3-1-4-8-17)21(13-19)24-16-18-9-5-2-6-10-18/h1-14H,15-16H2
InChI key
XDDLXZHBWVFPRG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Giovanna Baron et al.
Biochemical pharmacology, 173, 113726-113726 (2019-11-30)
The aim of this work was to profile, by using an HPLC-MS/MS method, cranberry compounds and metabolites found in human urine after ingestion of a highly standardized cranberry extract (Anthocran®). Two different strategies were adopted for the data analysis: a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service