Skip to Content
Merck
All Photos(3)

Key Documents

D23807

Sigma-Aldrich

1,3-Diaminopropane dihydrochloride

98%

Synonym(s):

1,3-Propanediamine, 1,3-Propanediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)3NH2·2HCl
CAS Number:
Molecular Weight:
147.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

246-250 °C (lit.)

SMILES string

Cl[H].Cl[H].NCCCN

InChI

1S/C3H10N2.2ClH/c4-2-1-3-5;;/h1-5H2;2*1H

InChI key

HYOCSVGEQMCOGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Tetradentate bis-phosphine ligands (P(2)N(2) and P(2)S(2)) and their Rh(III), Ni(II) and (105)Rh complexes: X-ray crystal structures of trans-[RhCl(2)(L2)]PF(6), [Ni(L2)](PF(6))(2) and μ-O(2)SO(2)-[Ni(L5)](2)(PF(6))(2).: This study explores the synthesis and characterization of tetradentate bis-phosphine ligands and their complexes, demonstrating significant advancements in ligand design and potential applications in catalysis and medicinal chemistry (Cagnolini et al., 2011).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nidhi Mishra et al.
European journal of medicinal chemistry, 43(7), 1530-1535 (2007-11-06)
The synthesis of novel 1,3-diaryl propenone derivatives and their antimalarial activity in vitro against asexual blood stages of human malaria parasite, Plasmodium falciparum, are described. Chalcone derivatives were prepared via Claisen-Schmidt condensation of substituted aldehydes with substituted methyl ketones. Antiplasmodial
Iwao Suzuki et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(5), 655-657 (2006-06-14)
Pyrene-appended alpha-cyclodextrin (3) in which a trimethylenediamine linker connected the pyrene residue to the alpha-cyclodextrin moiety showed pH-dependent fluorescence intensity changes. The fluorescence intensity was almost linearly changed within the pH range of 5 - 10. The unique fluorescence response
Rolando Barbucci et al.
Acta biomaterialia, 8(12), 4244-4252 (2012-09-18)
Hybrid magnetic hydrogels are of interest for applications in biomedical science as controlled drug-delivery systems. We have developed a strategy to obtain novel hybrid hydrogels with magnetic nanoparticles (NPs) of CoFe(2)O(3) and Fe(3)O(4) as crosslinker agents of carboxymethylcellulose (CMC) or
Jorge Martín et al.
Applied and environmental microbiology, 77(16), 5688-5696 (2011-07-05)
Filamentous fungi produce an impressive variety of secondary metabolites; many of them have important biological activities. The biosynthesis of these secondary metabolites is frequently induced by plant-derived external elicitors and appears to also be regulated by internal inducers, which may
Roland Meier et al.
Chemical communications (Cambridge, England), (38)(38), 3960-3962 (2007-09-27)
The equilibrium between the twist-boat (tb) and half-chair (hc) conformers of the central diamine chelate ring of [Fe(III)(tmdta)]- in solids and aqueous solution has been studied by Raman spectroscopy, supported by calculated Raman spectra using Density Functional Theory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service