Skip to Content
Merck
All Photos(2)

Key Documents

D221902

Sigma-Aldrich

(2-Dodecen-1-yl)succinic anhydride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26O3
CAS Number:
Molecular Weight:
266.38
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

9.2 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

95%

form

solid

bp

180-182 °C/5 mmHg (lit.)

mp

41-43 °C (lit.)

SMILES string

CCCCCCCCC\C=C/CC1CC(=O)OC1=O

InChI

1S/C16H26O3/c1-2-3-4-5-6-7-8-9-10-11-12-14-13-15(17)19-16(14)18/h10-11,14H,2-9,12-13H2,1H3/b11-10+

InChI key

UYCICMIUKYEYEU-ZHACJKMWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Nakamura et al.
The Journal of toxicological sciences, 24(2), 123-131 (1999-06-01)
The sensitivities of three skin sensitization tests such as the Guinea pig maximization test (GPMT), Adjuvant and patch test (APT) and Buehler test (BT), were quantitatively compared with reference to induction and challenge concentrations. Four chemical which had different physico-chemical
Vinod Vellora Thekkae Padil et al.
Journal of agricultural and food chemistry, 63(14), 3757-3765 (2015-03-24)
Esterifications of the tree-based gum, gum karaya (GK), using dodecenylsuccinic anhydride (DDSA) were carried out in aqueous solutions. GK was deacetylated using alkali treatment to obtain deacetylated gum karaya (DGK). The DGK and its DDSA derivative were characterized using gel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service