CLS1705100
Corning® coliform water test sample container, sterile
without sodium thiosulfate tablet, capacity 100-120 mL
Synonym(s):
Coliform Water Test Container, Sterile Water Sample Container, Water Test Sample Container
All Photos(1)
About This Item
UNSPSC Code:
41161502
NACRES:
NB.13
Recommended Products
manufacturer/tradename
Corning 1705-100
availability
not available in EU
capacity
100-120 mL
Related Categories
General description
- Well defined EPA Fill Line for 100 mL
- Reinforced custody string
- Large thumb tabs to reduce contamination and ease for opening
- 2 thumb taps on either side of the locking mechanism
- 1 in. headspace between water level and underside of closed lid
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)