Skip to Content
Merck
All Photos(1)

Key Documents

CHP-507

Sigma-Aldrich

Saturated Phosphatidylcholine Liposomes

100% 20:0 PC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

Pricing and availability is not currently available.

Quality Level

composition

Phosphate buffered saline

concentration

10 mM

impurities

100 mol % 20:0 1,2-diarachidoyl-sn-glycero-3-phosphocholine

particle size

100 nm

pH

7.4

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CHP-521CHP-520CHP-502
composition

Phosphate buffered saline

composition

Phosphate buffered saline

composition

Phosphate buffered saline

composition

Phosphate buffered saline

concentration

10 mM

concentration

10 mM

concentration

10 mM

concentration

10 mM

impurities

100 mol % 20:0 1,2-diarachidoyl-sn-glycero-3-phosphocholine

impurities

30 mol % Cholesterol, 70 mol % 19:0 1,2-dinonadecanoyl-sn-glycero-3-phosphocholine

impurities

30 mol % Cholesterol, 70 mol % 18:0 1,2-distearoyl-sn-glycero-3-phosphocholine (DSPC)

impurities

100 mol % 15:0 1,2-dipentadecanoyl-sn-glycero-3-phosphocholine

particle size

100 nm

particle size

100 nm

particle size

100 nm

particle size

100 nm

pH

7.4

pH

7.4

pH

7.4

pH

7.4

General description

Liposomes are extensively used to study the interaction of proteins, peptides and other molecules with the surface of a lipid membrane. Liposomes made from saturated PC lipids have a high liquid to gel phase transition temperature and have different fatty acid lengths from 12:0-24:0 with and without cholesterol.

Application

Drug delivery
Lipid-protein interactions

Storage and Stability

Liposomes should never be frozen. Liposomes should be stored in the dark at 4°C, except when brought to room temperature for brief periods prior to use.

Liposomes are made under sterile conditions. If you need to take multiple aliquots out of the vial, it is advised to take extreme care in not contaminating the vial. It is recommended to handle the vial under a sterile hood to maintain the sterility of the product. Liposomes should never be frozen. Ice crystals that form during freezing will rupture the lipid membrane of the liposomes and change the size of liposomes particles.

Legal Information

Cellsome is a trademark of Encapsula NanoSciences
Product of Encapsula Nanosciences

Disclaimer

For research use only

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service