Recommended Products
description
AldrichCPR
form
solid
SMILES string
C12=CC=CN=C1N=CC=C2
InChI
1S/C8H6N2/c1-3-7-4-2-6-10-8(7)9-5-1/h1-6H
InChI key
FLBAYUMRQUHISI-UHFFFAOYSA-N
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Accounts of chemical research, 53(9), 1944-1956 (2020-09-04)
Bimetallic motifs are a structural feature common to some of the most effective and synthetically useful catalysts known, including in the active sites of many metalloenzymes and on the surfaces of industrially relevant heterogeneous materials. However, the complexity of these
Chemistry, an Asian journal, 15(2), 279-286 (2019-12-04)
For a long time, the cobalt(II) complex ([Co(napy)4 ](ClO4 )2 ) (napy=1, 8-naphthyridine) has been considered as an eight-coordinate complex without any structural proof. After careful considerations, two complexes [Co(napy)2 Cl2 ] (1) and [Co(napy)4 ](ClO4 )2 (2) based on
Biochemical and biophysical research communications, 491(2), 416-422 (2017-07-20)
Nicotinamide N-methyltransferase (NNMT) is a S-adenosyl-l-methionine (SAM)-dependent enzyme that catalyzes N-methylation of nicotinamide (NA) and other pyridines to form N-methyl pyridinium ions. Here we report the first ternary complex X-ray crystal structures of monkey NNMT and mouse NNMT in bound
Journal of chemical information and modeling, 60(11), 5487-5498 (2020-05-19)
The urgent need for new treatments for the chronic lung disease idiopathic pulmonary fibrosis (IPF) motivates research into antagonists of the RGD binding integrin αvβ6, a protein linked to the initiation and progression of the disease. Molecular dynamics (MD) simulations
Neuropharmacology, 158, 107731-107731 (2019-08-04)
Disruption of the hypothalamic-pituitary-adrenal axis is an established finding in patients with anxiety and/or depression. Chronic corticosterone administration in animals has been proposed as a model for the study of these stress-related disorders and the antidepressant action. Alterations of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service