Skip to Content
Merck
All Photos(1)

Key Documents

CDS020650

Sigma-Aldrich

Glycyl-L-histidyl-L-lysine

Synonym(s):

(2S)-6-Amino-2-{[(2S)-2-[(aminoacetyl)amino]-3-(1H-imidazol-4-yl)propanoyl]amino}hexanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24N6O4
Molecular Weight:
340.38
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

description

AldrichCPR

form

solid

SMILES string

O=C(N[C@H](C(O)=O)CCCCN)[C@@H](NC(CN)=O)CC1=CNC=N1

InChI

1S/C14H24N6O4/c15-4-2-1-3-10(14(23)24)20-13(22)11(19-12(21)6-16)5-9-7-17-8-18-9/h7-8,10-11H,1-6,15-16H2,(H,17,18)(H,19,21)(H,20,22)(H,23,24)/t10-,11-/m0/s1

InChI key

MVORZMQFXBLMHM-QWRGUYRKSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toyoshi Endo et al.
Endocrine-related cancer, 20(6), 767-776 (2013-09-10)
A variant located on 14q13.3 nearest to thyroid transcription factor-1 (TTF1) predisposes individuals to thyroid cancer, but whether this variant is related to the RET/PTC rearrangement associated with human papillary thyroid carcinomas (PTCs) is unknown. The aims of this study
Keith H Richards et al.
PloS one, 12(8), e0183482-e0183482 (2017-08-25)
Tyrosine and phenolic ring de-iodination of thyroid hormones (TH) is crucial for regulating their physiological activity. Furthermore, reactions such as de-carboxylation to thyronamines (TAM) and de-amination to thyroacetic acids (TAc) produce TH metabolites (THM) with distinct biological properties. This needs
T Di Palma et al.
Cell death & disease, 4, e729-e729 (2013-07-23)
The transcription factor Pax8, a member of the Paired-box gene family, is a critical regulator required for proper development and differentiation of thyroid follicular cells. Despite being Pax8 well characterized with respect to its role in regulating genes responsible for
Michela Vitiello et al.
Genes, 10(2) (2019-02-13)
PATZ1 is a transcriptional factor downregulated in thyroid cancer whose re-expression in thyroid cancer cells leads to a partial reversion of the malignant phenotype, including the capacity to proliferate, migrate, and undergo epithelial-to-mesenchymal transition. We have recently shown that PATZ1
Alessia Parascandolo et al.
Oncotarget, 9(4), 4496-4510 (2018-02-13)
Sonic Hedgehog (Shh) pathway regulates embryonic development of different organs including the thyroid gland. The aberrant activation of Shh signaling has been found in several types of cancer and according to recent evidences it represents an important regulator of tumor-stroma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service