Skip to Content
Merck
All Photos(1)

Key Documents

CDS010729

Sigma-Aldrich

perfluorooctanesulfonamide

AldrichCPR

Synonym(s):

1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-1-octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluorooctan-1-sulfonamide, Desethylsulfluramid, Perfluoroctylsulfonamide, Perfluorooctanesulfonamide, Perfluorooctanesulfonic acid amide, Perfluorooctylsulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H2F17NO2S
CAS Number:
Molecular Weight:
499.14
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

form

solid

SMILES string

NS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C8H2F17NO2S/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)29(26,27)28/h(H2,26,27,28)

InChI key

RRRXPPIDPYTNJG-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Coperchini et al.
Frontiers in endocrinology, 11, 612320-612320 (2021-02-06)
Per- and polyfluoroalkyl substances (PFAS) represent a group of synthetic compounds widely used in industry plants due to their low grade of degradation, surfactant properties, thermic and flame resistance. These characteristics are useful for the industrial production, however they are
Tang Jiawei et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 79(4), 731-740 (2019-04-13)
The potential negative effects of perfluoroalkyl substances (PFASs) discharged into aquatic environments are drawing increasing attention. However, little research has been undertaken on PFASs in wastewater from electroplating industrial parks. In this study, the concentration profiles and geographical distribution of
Boris Bugsel et al.
Analytical and bioanalytical chemistry, 412(20), 4797-4805 (2020-01-11)
The application of contaminated paper sludge on arable land in southwest Germany caused the occurrence of a broad range of poly- and perfluoroalkyl substances (PFASs) on soil. Recently, the dead-end transformation products (TPs) perfluorooctanoic acid and perfluorooctanesulfonic acid were detected in
Huangfang Xu et al.
Environment international, 143, 105952-105952 (2020-07-28)
Long-chain per- and polyfluoroalkyl substances (PFASs) and their short-chain alternatives have been produced and used extensively in China. However, it is unclear whether these compounds contribute to the risk of gestational diabetes mellitus (GDM) in women residing in contaminated areas.
Andrew Yeh et al.
Aquatic toxicology (Amsterdam, Netherlands), 190, 21-31 (2017-07-03)
We previously reported the bioaccumulation of contaminants of emerging concern (CECs), including pharmaceuticals and personal care products (PPCPs) and perfluorinated compounds, in field-collected juvenile Chinook salmon from urban estuaries of Puget Sound, WA (Meador et al., 2016). Although the toxicological

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service