CAD4855
Stainless steel 304 syringe needle
gauge 20, L 10 in.
All Photos(1)
About This Item
UNSPSC Code:
42142600
NACRES:
NB.03
Recommended Products
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science 4855
L
10 in.
gauge
20
Related Categories
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Standard Hub with 304 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)