Skip to Content
Merck
All Photos(3)

Key Documents

C75802

Sigma-Aldrich

Cholesteryl benzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H50O2
CAS Number:
Molecular Weight:
490.76
Beilstein:
3225829
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid crystal

optical activity

[α]20/D −15.0°, c = 2 in chloroform

mp

148-150 °C (lit.)

SMILES string

O=C(O[C@@H]1CC2=CC[C@]3([H])[C@@](CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CCCC(C)C)([H])[C@@]2(C)CC1)C5=CC=CC=C5

InChI

1S/C34H50O2/c1-23(2)10-9-11-24(3)29-16-17-30-28-15-14-26-22-27(36-32(35)25-12-7-6-8-13-25)18-20-33(26,4)31(28)19-21-34(29,30)5/h6-8,12-14,23-24,27-31H,9-11,15-22H2,1-5H3/t24-,27+,28+,29-,30+,31+,33+,34-/m1/s1

InChI key

UVZUFUGNHDDLRQ-LLHZKFLPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cholesteryl benzoate (CB) is a cholesteryl ester which is used in cholesteric liquid crystals by forming layered spiral staircases in which the rotation angle is temperature dependent.

Application

Cholesteryl benzoate (CB) is a structure directing agent which can be used in the growth of oriented lithium iron phosphate (LiFePO4) for the fabrication of lithium ion batteries. It can be used in the fabrication of polymer dispersed liquid crystals (PDLC) which show liquid crystallinity by cooling the material from the isotropic phase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility of cholesterol in bile salt-synthetic detergent mixed micelles & that of cholesteryl benzoate in synthetic detergent mixed micelles.
S Pal et al.
Indian journal of biochemistry & biophysics, 21(1), 17-22 (1984-02-01)
Thermal and optical characterization of polymer-dispersed liquid crystals.
Shanks RA and Staszczyk D
International Journal of Polymer Science, 2012(1), 116-120 (2012)
Zhen Huang et al.
International journal of pharmaceutics, 397(1-2), 130-135 (2010-07-27)
Evidences indicating the presence of phase transformations in the mixed cholesteryl benzoate (CBE) and cholesteryl butyrate (CBU) under the supercritical CO(2) pressurization, by means of differential scanning calorimetry (DSC) and X-ray diffraction (XRD), are presented in this work. These include
Solubility of cholesterol & cholesteryl benzoate in some polar & nonpolar organic solvents.
S Pal et al.
Indian journal of biochemistry & biophysics, 21(1), 12-16 (1984-02-01)
M L Das et al.
Indian journal of biochemistry & biophysics, 26(1), 24-29 (1989-02-01)
Phase behaviour and some physicochemical properties of cholesteryl benzoate-contained microemulsions of xylene and heptane with water using sodium deoxycholate and butan-1-ol as surfactant and co-surfactant respectively have been studied. While the microemulsion area of xylene depends insignificantly on the surfactant/co-surfactant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service